Learn More
ACROBiosystems Recombinant Protein;Human CD48 / SLAMF2 Protein, Fc Tag;HEK293;1MG;BC1,CD48,BCM1,SLAMF2,BLAST,BLAST1,MEM-102,TCT.1,BCM-1,SLAMF-2,BLAST-1
Catalog No. 501658244
Encompass_Preferred
Recombinant Protein;Human CD48 / SLAMF2 Protein, Fc Tag;1MG;Human CD48, Fc Tag (BC1-H5255) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Ser 220 (Accession # P09326-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 49.0 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human 2B4, His Tag (Cat. No. 2B4-H5224) at 2 μg/mL (100 μL/well) can bind Human CD48, Fc Tag (Cat. No. BC1-H5255) with a linear range of 0.078-2.5 μg/mL (QC tested).
Catalog No. 50-165-8244 Supplier ACROBiosystems Supplier No. BC1H52551MG
Recombinant Protein;Human CD48 / SLAMF2 Protein, Fc Tag;1MG;Human CD48, Fc Tag (BC1-H5255) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Ser 220 (Accession # P09326-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 49.0 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human 2B4, His Tag (Cat. No. 2B4-H5224) at 2 μg/mL (100 μL/well) can bind Human CD48, Fc Tag (Cat. No. BC1-H5255) with a linear range of 0.078-2.5 μg/mL (QC tested).