Learn More
ACROBiosystems Recombinant Protein;Human CD48 / SLAMF2 Protein;HEK293;100UG;BC1,CD48,BCM1,SLAMF2,BLAST,BLAST1,MEM-102,TCT.1,BCM-1,SLAMF-2,BLAST-1
Catalog No. 501658195
Encompass_Preferred
Recombinant Protein;Human CD48 / SLAMF2 Protein;100UG;Human CD48, His Tag (BC1-H5226) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Ser 220 (Accession # AAH16182).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.2 kDa. The protein migrates as 38-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human 2B4, Fc Tag (Cat. No. 2B4-H5255) at 5 μg/mL (100 μL/well) can bind Human CD48, His Tag (Cat. No. BC1-H5226) with a linear range of 0.156-2.5 μg/mL (QC tested).
Catalog No. 50-165-8195 Supplier ACROBiosystems Supplier No. BC1H5226100UG
Recombinant Protein;Human CD48 / SLAMF2 Protein;100UG;Human CD48, His Tag (BC1-H5226) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Ser 220 (Accession # AAH16182).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.2 kDa. The protein migrates as 38-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human 2B4, Fc Tag (Cat. No. 2B4-H5255) at 5 μg/mL (100 μL/well) can bind Human CD48, His Tag (Cat. No. BC1-H5226) with a linear range of 0.156-2.5 μg/mL (QC tested).