Learn More
ACROBiosystems Recombinant Protein;Human CD4 Protein, Fc Tag;HEK293;1MG;CD4,CD4,CD-4,CD4mut,CD-4mut,LEU3,LEU-3
Supplier:  ACROBiosystems CD4H52591MG
Encompass_Preferred
Recombinant Protein;Human CD4 Protein, Fc Tag;1MG;Human CD4, Fc Tag (CD4-H5259) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Pro 396 (Accession # AAH25782).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 67.5 kDa. The protein migrates as 76 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized HIV-1 GP120 (CN54), His Tag (Cat. No. GP4-V15227) at 5μg/mL (100 μL/well) can bind Human CD4, Fc Tag (Cat. No. CD4-H5259) with a linear range of 4.9-78 ng/mL (QC tested).
Catalog No. 50-165-7475