Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human CD4, Fc Tag & Avi Tag (Avitag™);HEK293;200UG;CD4,CD4,CD-4,CD4mut,CD-4mut,LEU3,LEU-3
Catalog No. 501658293
Encompass_Preferred
Recombinant Protein;Biotinylated Human CD4, Fc Tag & Avi Tag (Avitag™);200UG;Biotinylated Human CD4, Fc Tag (CD4-H82F3) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Pro 396 (Accession # P01730-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 69.7 kDa. As a result of glycosylation, the protein migrates as 80 kDa under reducing (R) condition, and 150 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized HIV GP120 (HIV), His Tag (Cat. No. GP5-V15224) at 5 μg/mL (100 μL/well) can bind Biotinylated Human CD4, Fc Tag (Cat. No. CD4-H82F3) with a linear range of 0.5-8 ng/mL (QC tested).
Catalog No. 50-165-8293 Supplier ACROBiosystems Supplier No. CD4H82F3200UG
Recombinant Protein;Biotinylated Human CD4, Fc Tag & Avi Tag (Avitag™);200UG;Biotinylated Human CD4, Fc Tag (CD4-H82F3) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Pro 396 (Accession # P01730-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 69.7 kDa. As a result of glycosylation, the protein migrates as 80 kDa under reducing (R) condition, and 150 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized HIV GP120 (HIV), His Tag (Cat. No. GP5-V15224) at 5 μg/mL (100 μL/well) can bind Biotinylated Human CD4, Fc Tag (Cat. No. CD4-H82F3) with a linear range of 0.5-8 ng/mL (QC tested).