Learn More
ACROBiosystems Recombinant Protein;Rhesus macaque CD4 Protein;HEK293;1MG;CD4,CD4,CD-4,CD4mut,CD-4mut,LEU3,LEU-3
Catalog No. 501658599
Encompass_Preferred
Recombinant Protein;Rhesus macaque CD4 Protein;1MG;Rhesus macaque CD4, His Tag (CD4-C52H7) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Trp 390 (Accession # G7N5T8).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 42.3 kDa. The protein migrates as 52-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8599 Supplier ACROBiosystems Supplier No. CD4C52H71MG
Recombinant Protein;Rhesus macaque CD4 Protein;1MG;Rhesus macaque CD4, His Tag (CD4-C52H7) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Trp 390 (Accession # G7N5T8).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 42.3 kDa. The protein migrates as 52-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.