Learn More
ACROBiosystems Recombinant Protein;Human CD4 Protein;HEK293;1MG;LE3,CD4,CD-4,CD4mut,CD-4mut,LEU3,LEU-3
Catalog No. 501658539
Encompass_Preferred
Recombinant Protein;Human CD4 Protein;1MG;Human CD4, His Tag (LE3-H5228) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Trp 390 (Accession # AAH25782).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 41.5 kDa. The protein migrates as 47-54 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD4, His Tag (Cat. No. LE3-H5228) at 5 μg/mL (100 μL/well) can bind Biotinylated Human GP120 (HIV) (Cat. No. GP5-V182E9) with a linear range of 0.039-0.625 μg/mL (QC tested).
Catalog No. 50-165-8539 Supplier ACROBiosystems Supplier No. LE3H52281MG
Recombinant Protein;Human CD4 Protein;1MG;Human CD4, His Tag (LE3-H5228) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Trp 390 (Accession # AAH25782).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 41.5 kDa. The protein migrates as 47-54 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD4, His Tag (Cat. No. LE3-H5228) at 5 μg/mL (100 μL/well) can bind Biotinylated Human GP120 (HIV) (Cat. No. GP5-V182E9) with a linear range of 0.039-0.625 μg/mL (QC tested).