Learn More
ACROBiosystems Recombinant Protein;Human CD4 Protein;HEK293;200UG;LE3,CD4,CD-4,CD4mut,CD-4mut,LEU3,LEU-3
Supplier: ACROBiosystems LE3H5228200UG
Recombinant Protein;Human CD4 Protein;200UG;Human CD4, His Tag (LE3-H5228) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Trp 390 (Accession # AAH25782).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 41.5 kDa. The protein migrates as 47-54 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD4, His Tag (Cat. No. LE3-H5228) at 5 μg/mL (100 μL/well) can bind Biotinylated Human GP120 (HIV) (Cat. No. GP5-V182E9) with a linear range of 0.039-0.625 μg/mL (QC tested).