Learn More
Diagnostic Biosystems CD5 CD5 (CD5/54/F6)
Catalog No. 502407478
Encompass_Preferred
This antibody is Recognizes a 67kDa transmembrane protein, which is identified as CD5. The CD5 antigen is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, Important in identification of T cells and most T cell lymphomas, some low-grade B cell lymphomas, thymic carcinoma.
Catalog No. 50-240-7478 Supplier Diagnostic Biosystems Supplier No. PDM589
This antibody is Recognizes a 67kDa transmembrane protein, which is identified as CD5. The CD5 antigen is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, Important in identification of T cells and most T cell lymphomas, some low-grade B cell lymphomas, thymic carcinoma.