Learn More
Biogenex Laboratories CD5 : 4C7 : 6mL
SDP
Supplier:  Biogenex Laboratories AM4305M
Encompass_Preferred
The CD5 antigen, also known as T1, is a 67 kD single chain glycoprotein expressed on normal and malignant T cells and on chronic lymphocytic leukemia cells. It is found in high density on medullary thymocytes and in low density on cortical thymocytes.
Catalog No. 50-197-8768