Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 921MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: C5/473. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: 293T, Ramos, or MOLT-4 Cells. Tonsil. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Human CD5 recombinant protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 67kDa.Chromosome: 11q12.2. Gene Symbol: CD5. Gene ID: 921. Ensemble: ENSG00000110448. Uniprot: NP_055022.
Catalog No. 50-188-5324