Learn More
Medchemexpress LLC CD5 Human HEK293 100ug
SDP
Catalog No. 5000182795
Encompass_Preferred
The CD5 protein is a potential receptor that regulates T cell proliferation Its interactions with CD72/LYB-2 and PTPN6/SHP-1 indicate multifaceted roles in cellular processes serving as key mediators of T cell responses CD5 Protein Human (HEK293 His) is the recombinant human-derived CD5 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-82795 Supplier Medchemexpress LLC Supplier No. HYP72920100UG
The CD5 protein is a potential receptor that regulates T cell proliferation Its interactions with CD72/LYB-2 and PTPN6/SHP-1 indicate multifaceted roles in cellular processes serving as key mediators of T cell responses CD5 Protein Human (HEK293 His) is the recombinant human-derived CD5 protein expressed by HEK293 with C-His labeled tag