Learn More
Sino Biological CD5 / LEU1 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 11027MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD5 (rh CD5; Catalog#11027-H08H; NP_055022.2; Met 1-Pro 372). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-7548