Learn More
Biocare Medical, LLC CD5 Antibody - Concentrate, 1.0 ml
SDP
Supplier:  Biocare Medical, LLC CM099C
Encompass_Preferred
CD5 antibody is a T-cell associated marker that is also expressed by two B-cell neoplasms: lymphocytic leukemia and mantle cell lymphoma. CD5 antigen is expressed in 95% of thymocytes and 72% of peripheral blood lymphocytes. It has been shown to react with thymic carcinomas, but rarely in thymomas. It has also been observed in a subset of intravascular large B-cell lymphomas and marks some anaplastic large cell lymphomas. CD5 has been reported to be very useful in marking mantle cell lymphoma when used in tandem with CD23, cyclin D1, and CD10 (CD10 is negative; CD5 and cyclin D1 are positive for mantle cell lymphoma).
Catalog No. 50-825-08