Learn More
Medchemexpress LLC CD5 Mouse HEK293 50ug
SDP
Catalog No. 5000182442
Encompass_Preferred
CD5 protein acting as a receptor crucially regulates T-cell proliferation by engaging in vital interactions with CD72/LYB-2 Additionally its potential interactions with PTPN6/SHP-1 highlight its integral role in essential signaling pathways CD5 Protein Mouse (HEK293 His) is the recombinant mouse-derived CD5 protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-82442 Supplier Medchemexpress LLC Supplier No. HYP7272250UG
CD5 protein acting as a receptor crucially regulates T-cell proliferation by engaging in vital interactions with CD72/LYB-2 Additionally its potential interactions with PTPN6/SHP-1 highlight its integral role in essential signaling pathways CD5 Protein Mouse (HEK293 His) is the recombinant mouse-derived CD5 protein expressed by HEK293 with C-6 His labeled tag