Learn More
Beckman Coulter Beckman Coulter CD5 Pacific Blue Antibody, Clone BL1a, 0.5 mL (A82790)
Supplier: Beckman Coulter A82790
Monoclonal antibody for detection of human CD5 by flow cytometry. Conjugated to Pacific Blue and supplied as a 0.5 mL liquid in ASR format; clone BL1a provides reliable staining of CD5-positive lymphocyte populations for immunophenotyping applications.
- Target: CD5 (human).
- Clone: BL1a.
- Conjugate: Pacific Blue (PB).
- Format: Liquid, ASR.
- Volume: 0.5 mL.
- Application: Flow cytometry (cell surface staining).
- Typical use: Immunophenotyping of T and B lymphocyte subsets.
Description
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.