Learn More
Medchemexpress LLC CD5 Rat HEK293 Hi 5ug
SDP
Catalog No. 5000178558
Encompass_Preferred
CD5 protein a pivotal T-cell receptor regulates T-cell proliferation through interactions with CD72/LYB-2 Its potential interaction with PTPN6/SHP-1 underscores its involvement in critical signaling pathways CD5 Protein Rat (HEK293 His) is the recombinant rat-derived CD5 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-78558 Supplier Medchemexpress LLC Supplier No. HYP742835UG
CD5 protein a pivotal T-cell receptor regulates T-cell proliferation through interactions with CD72/LYB-2 Its potential interaction with PTPN6/SHP-1 underscores its involvement in critical signaling pathways CD5 Protein Rat (HEK293 His) is the recombinant rat-derived CD5 protein expressed by HEK293 with C-His labeled tag