Learn More
Enzo Life Sciences CD5 Recombinant monoclonal antibody (RM314) (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS6720100
Encompass_Preferred
Clone: RM314. Isotype: IgG. Immunogen: Synthetic peptide corresponding to the C-terminus of human CD5. Source: Purified from animal origin-free supernatant. Species reactivity: Human. Applications: IHC (PS), WB. Purity: Protein A affinity purified. Formulation: Liquid. In 1x PBS containing 50% glycerol, 1% BSA and 0.09% sodium azide.
Catalog No. 50-201-3200