Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3385MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Bioactive, Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: 186-2G9. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: HL-60 or THP-1 cells. Lymph node, tonsil. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Stimulated human leukocytes. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 110-160kDa.Chromosome: 19p13.2. Gene Symbol: ICAM3. Gene ID: 3385. Ensemble: ENSG00000076662. Uniprot: NP_002153.
Catalog No. 50-188-4792