Learn More
Biotium CD50 / ICAM3(ICO-60), CF647 conjugate, 0.1mg/mL
Supplier:  Biotium BNC471040500
Encompass_Preferred
Recognizes the N-glycosylated glycoprotein of 120 kDa with intra-chain disulfide bonds, identified as CD50 or ICAM-3. CD50 is the major ligand for LFA-1 (CD11a/CD18) and may have signaling role to increase adhesion. It is expressed on thymocytes and T lymphocytes and is resistant to treatment with phosphatidylinositol (PI) phospholipase C.
Catalog No. 50-006-21177