Learn More
Enzo Life Sciences CD51 (human) monoclonal antibody (L230) (100 µg)
SDP
Supplier:  Enzo Life Sciences ALX803304C100
Encompass_Preferred
Clone: L230. Host: Mouse. Isotype: IgG1. Immunogen: Human endothelial cells. UniProt: P06756. Species Reactivity: Human. Specificity: Recognizes heterodimers αVβ3, αVβ1, αVβ5 and αVβ6. Crossreactivity: Does not cross-react with other species. Applications: ELISA. Flow Cytometry, IF, IHC, IP, FUNC. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS containing 0.05% sodium azide. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C.
Catalog No. NC0894051