Learn More
Genscript Corporation CD52 hFc Chimera Mouse
Catalog No. 5000056935
Encompass
CD52 also known as CAMPATH-1 antigen HE5 and gp20 is a cell surface glycoprotein that can be targeted to induce immune suppression by complement-mediated cell lysis Mature human CD52 is a 12 amino acid peptide that is tethered to the cell surface with a GPI linkage CD52 may play a role in carrying and orienting carbohydrate as well as having a more specific role
Catalog No. 50-000-56935 Supplier Genscript Corporation Supplier No. Z04324500
CD52 also known as CAMPATH-1 antigen HE5 and gp20 is a cell surface glycoprotein that can be targeted to induce immune suppression by complement-mediated cell lysis Mature human CD52 is a 12 amino acid peptide that is tethered to the cell surface with a GPI linkage CD52 may play a role in carrying and orienting carbohydrate as well as having a more specific role