Learn More
ACROBiosystems ACROBiosystems Biotinylated Human CD52 Protein, His,Avitag™
Supplier:  ACROBiosystems CD2H82E8200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human CD52, His,Avitag (CD2-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ser 36 (Accession # P31358-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 13.5 kDa. The protein migrates as 16 kDa and 17-22 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Monoclonal Anti-Human CD52 Antibody, Human IgG1 at 10 μg/mL (100 μL/well) can bind Biotinylated Human CD52, His,Avitag (Cat. No. CD2-H82E8) with a linear range of 0.005-0.625 μg/mL (QC tested).;Anti-Human CD52 MAb (Human IgG1) captured on CM5 chip via anti-human IgG Fc antibodies surface can bind Biotinylated Human CD52, His,Avitag (Cat. No. CD2-H82E8) with an affinity constant of 71.8 nM as determined in a SPR assay (Biacore T200) (Routinely tested).
Catalog No. 50-195-6459