Learn More
Enzo Life Sciences CD52 Recombinant monoclonal antibody (YTH 34.5-G2b) (Rat IgG2b kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4550200
Encompass_Preferred
Clone: YTH 34.5-G2b (Campath-1G). Isotype: IgG2bκ. Immunogen: Human peripheral mononuclear cells enriched for T-cells. Species reactivity: Human Rhesus monkey. Applications: ELISA, Flow Cytometry, IHC (FS), IHC (PS), WB in vivo Assay, Cytotoxicity studies. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-2982