Learn More
ACROBiosystems Recombinant Protein;Human CD52 Protein, Fc Tag;HEK293;100UG;CD2,CDW52,HE5,CD52,CAMPATH-1 antigen
Supplier:  ACROBiosystems CD2H525A100UG
Encompass_Preferred
Recombinant Protein;Human CD52 Protein, Fc Tag;100UG;Human CD52, Fc Tag (CD2-H525a) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ser 36 (Accession # P31358-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 27.9 kDa. The protein migrates as 38-43 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD52, Fc Tag (Cat. No. CD2-H525a) at 10 μg/mL (100 μL/well) can bind Alemtuzumab with a linear range of 0.313-10 μg/mL (QC tested).
Catalog No. 50-165-8038