Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human CD52, Fc Tag, Avi Tag (Avitag™);HEK293;25UG;CD2,CDW52,HE5,CD52,CAMPATH-1 antigen
Supplier:  ACROBiosystems CD2H82F325UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human CD52, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human CD52, Fc Tag (CD2-H82F3) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ser 36 (Accession # P31358-1 ).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 29.7 kDa. As a result of glycosylation, the protein migrates as 38-45 kDa under reducing (R) condition, and 66-100 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Anti CD52 MAb, Human IgG1 at 10 μg/mL (100 μL/well) can bind Biotinylated Human CD52, Fc Tag (Cat. No. CD2-H82F3) with a linear range of 0.625-10 μg/mL (QC tested).
Catalog No. 50-165-8387