Learn More
Enzo Life Sciences CD54 (human) (domain D1) monoclonal antibody (RR1/1) (50µg)
SDP
Supplier:  Enzo Life Sciences ALX805016C050
Encompass_Preferred
Clone: RR1/1. Isotype: IgG1. Immunogen: Human CD54 (ICAM-1). Species reactivity: Human. Specificity: Recognizes the D1 domain of CD54. Applications: ELISA, Flow Cytometry, IHC (FS), WB, FUNC. Formulation: Liquid. In PBS, pH 7.2-7.4.
Catalog No. 50-201-0032