Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology CD54/ICAM-1 Monoclonal Antibody, Mouse, Human, 100 μl
SDP

Catalog No. 502732525
Encompass

This monoclonal antibody is designed for the detection of endogenous levels of total CD54/ICAM-1 protein. It is produced by immunizing animals with SDS-denatured, formalin-fixed cultured COLO-38 human melanoma cells. CD54/ICAM-1 is a cell surface glycoprotein belonging to the immunoglobulin superfamily that plays a vital role in leukocyte trafficking, host defense, and cell signaling pathways, including the activation of Src family kinases.

  • Host species: Mouse
  • Isotype: IgG1
  • Validated applications: Immunofluorescence (immunocytochemistry) and flow cytometry (fixed/permeabilized and live)
  • Species reactivity: Human
  • Storage conditions: -20°C
  • Formulation: Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/ml BSA, 50% glycerol and less than 0.02% sodium azide

Catalog No. 50-273-2525 Supplier Cell Signaling Technology Supplier No. 62133S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This monoclonal antibody is designed for the detection of endogenous levels of total CD54/ICAM-1 protein. It is produced by immunizing animals with SDS-denatured, formalin-fixed cultured COLO-38 human melanoma cells. CD54/ICAM-1 is a cell surface glycoprotein belonging to the immunoglobulin superfamily that plays a vital role in leukocyte trafficking, host defense, and cell signaling pathways, including the activation of Src family kinases.

  • Host species: Mouse
  • Isotype: IgG1
  • Validated applications: Immunofluorescence (immunocytochemistry) and flow cytometry (fixed/permeabilized and live)
  • Species reactivity: Human
  • Storage conditions: -20°C
  • Formulation: Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/ml BSA, 50% glycerol and less than 0.02% sodium azide

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.