Learn More
Genscript Corporation CD55 His Cynomolgus
Supplier:  Genscript Corporation Z04124100
Encompass
Decay Accelerating Factor (or CD55) is a major regulator of the alternative and classical pathways of complement activation and is expressed on all serum-exposed cells It is commonly hijacked by invading pathogens including many enteroviruses and uropathogenic Escherichia coli to promote cellular attachment prior to infection
Catalog No. 50-000-58462