Learn More
Enzo Life Sciences CD55 Recombinant monoclonal antibody (LU30) (Mouse IgG1 lambda) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6290200
Encompass_Preferred
Clone: LU30. Isotype: IgG1λ. Immunogen: Human CD55. Species reactivity: Human. Specificity: The antibody binds to CD55 with a Kd of 13 nM. Applications: ELISA, IHC, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3153