Learn More
ACROBiosystems Recombinant Protein;Human CD55 / DAF Protein;HEK293;100UG;CD5,CD55,DAF,CR,CROM,TC
Catalog No. 501657727
Encompass_Preferred
Recombinant Protein;Human CD55 / DAF Protein;100UG;Human CD55, His Tag (CD5-H5225) is expressed from human 293 cells (HEK293). It contains AA Asp 35 - Ser 353 (Accession # NP_000565.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 35.8 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7727 Supplier ACROBiosystems Supplier No. CD5H5225100UG
Recombinant Protein;Human CD55 / DAF Protein;100UG;Human CD55, His Tag (CD5-H5225) is expressed from human 293 cells (HEK293). It contains AA Asp 35 - Ser 353 (Accession # NP_000565.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 35.8 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.