Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4684MSM1XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM128. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Cerebellum, Pancreas, Neuroblastoma. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Membrane preparation of a small cell lung carcinoma. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 180, 145, 125kDa.Chromosome: 11q23.1. Gene Symbol: NCAM1. Gene ID: 4684. Ensemble: ENSG00000149294. Uniprot: NP_000606.
Catalog No. 50-188-4997