Learn More
CAPRICO BIOTECHNOLOGIES INC CD56 Antibody APC-Cyanine7, 4106095
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4106095
Encompass_Preferred
The CD56 (NCAM) Antibody (APC-Cyanine7) from Caprico Biotechnologies is an antibody that targets CD56, also known as neural cell adhesion molecule (NCAM). It is conjugated to APC-Cyanine7 and is suitable for flow cytometry, immunofluorescence microscopy and immunohistochemistry assays. This mouse monoclonal antibody recognizes the 140-200 kDa CD56 antigen and has been tested on human samples. According to the product details, it shows no cross-reactivity with other human antigens. The antibody is provided at 0.1 mg in PBS with 0.1% sodium azide and 0.2% gelatin.
Catalog No. 50-227-9206