Learn More
CAPRICO BIOTECHNOLOGIES INC CD56 APC/IFluor700
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4106175
Encompass_Preferred
The CD56 (APC-iFluor700) antibody from Caprico Biotechnologies is a mouse monoclonal antibody that recognizes the CD56 antigen, also known as neural cell adhesion molecule (NCAM). It is conjugated to APC-iFluor700 and is intended for flow cytometry applications. CD56 is expressed on NK cells and a subset of T cells. This antibody can be used for immunophenotyping and analyzing these cell populations by flow cytometry. The APC-iFluor700 conjugate provides bright fluorescence and high photostability. This antibody is supplied as 100 tests in PBS with 0.1% sodium azide and 0.2% BSA.
Catalog No. 50-227-9211