Learn More
CAPRICO BIOTECHNOLOGIES INC Anti-CD56 Antibody, iFluor 647, 4106125
Catalog No. 502279212
Encompass_Preferred
The CapricoBio Anti-CD56 (NCAM) Antibody [4106125] is a mouse monoclonal IgG2b antibody that has been conjugated to iFluor 647. It is specific for human CD56, also known as NCAM (neural cell adhesion molecule), with no cross-reactivity to other human proteins. CD56 is expressed on NK cells and a subset of T cells. This antibody can be used for flow cytometry, immunofluorescence microscopy, and other fluorescent detection methods. It is supplied as 100 tests in PBS with 0.1% sodium azide and 0.2% BSA.
This item is not returnable. View return policy
Catalog No. 50-227-9212 Supplier CAPRICO BIOTECHNOLOGIES INC Supplier No. 4106125
The CapricoBio Anti-CD56 (NCAM) Antibody [4106125] is a mouse monoclonal IgG2b antibody that has been conjugated to iFluor 647. It is specific for human CD56, also known as NCAM (neural cell adhesion molecule), with no cross-reactivity to other human proteins. CD56 is expressed on NK cells and a subset of T cells. This antibody can be used for flow cytometry, immunofluorescence microscopy, and other fluorescent detection methods. It is supplied as 100 tests in PBS with 0.1% sodium azide and 0.2% BSA.