Learn More
CAPRICO BIOTECHNOLOGIES INC CD56 (PE/CF594) Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4106205
Encompass_Preferred
The Caprico CD56 (PE-CF594) antibody is a mouse monoclonal antibody that specifically binds to human CD56. It is conjugated to PE-CF594 fluorescent dye and is intended for use in flow cytometry to identify natural killer cells. This antibody is supplied as a liquid in PBS with 0.1% sodium azide and 1% BSA. The concentration is 0.5 mg/mL and the clone number is MEM-188. The immunogen is an NK leukemia cell line. This antibody can be used for research purposes to study NK cells.
Catalog No. 50-227-9205