Learn More
CAPRICO BIOTECHNOLOGIES INC CD56 Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4192075
Encompass_Preferred
CD56 Antibody from is a mouse monoclonal IgG1 antibody that recognizes human CD56. It is unconjugated and has been tested by flow cytometry. The antibody reacts with natural killer cells and a subset of T cells. It has a concentration of 0.5 mg/mL and is supplied in PBS with 0.1% sodium azide. The recommended usage is 5 μL per test. It has a 12 month shelf life when stored at 2-8°C. This antibody can be used for applications such as flow cytometry, immunohistochemistry, and cell and tissue staining.
Catalog No. 50-227-9216