Learn More
CAPRICO BIOTECHNOLOGIES INC CD56 PE-Cyanine5, 4106075
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4106075
Encompass_Preferred
The Caprico CD56 (PE-Cyanine5) antibody is a mouse monoclonal antibody that recognizes the CD56 antigen, also known as neural cell adhesion molecule (NCAM). It is conjugated to PE-Cyanine5 and is intended for use in flow cytometry. This antibody stains natural killer cells and a subset of T cells. It is useful for studying natural killer cell biology and for identifying natural killer cell neoplasms. The antibody is available in sizes ranging from 25 tests to 1000 tests.
Catalog No. 50-227-9209