Learn More
Biocare Medical, LLC CD57 (Natural Killer Cell) Antibody - Concentrate, 1.0 ml
SDP
Supplier:  Biocare Medical, LLC CM007C
Encompass_Preferred
CD57 antibody [NK-1] marks a subset of lymphocytes known as natural killer (NK) cells. Follicular center cell lymphomas often contain many NK cells within the neoplastic follicles. Studies have shown that CD57 expression is present in normal and neoplastic pituitaries. It has been reported that CD57 reactivity may be used as an additional immunophenotypic criterion in distinguishing nodular lymphocyte predominance Hodgkin disease from nodular sclerosing Hodgkin disease, T-cell-rich B-cell lymphoma, and follicular lymphoma. CD57 [NK-1] also reportedly stains neuroendocrine cells and their respective tumors.
Catalog No. 50-824-17