Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 27087MSM1XP1
Encompass_Preferred
Product Type: Antibody. Application: IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM527. Clonality: Monoclonal. Host: Mouse. Isotype: IgM kappa. Positive Control: Lymph node or Tonsil. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Human peripheral blood mononuclear cells. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: ~110kDa.Chromosome: 11q25. Gene Symbol: B3GAT1. Gene ID: 27087. Ensemble: ENSG00000109956. Uniprot: NP_061114.
Catalog No. 50-188-4737