Learn More
CAPRICO BIOTECHNOLOGIES INC CD57 Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4033075
Encompass_Preferred
The CD57 Antibody is a mouse monoclonal antibody that recognizes human CD57. CD57 is expressed on subsets of NK cells, T cells, and monocytes. This antibody is unconjugated and is recommended for detection of CD57 protein in flow cytometry, immunohistochemistry, and Western blot applications. It is supplied as 100 μg purified antibody in PBS with 0.09% sodium azide and 0.2% gelatin.
Catalog No. 50-227-9219