Learn More
Biolegend Brilliant Violet 785™ anti-human CD57 Recombinant
Supplier:  Biolegend 393330
Encompass_Preferred
Brilliant Violet 785™ anti-human CD57 Recombinant [QA17A04]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1880830