Learn More
CAPRICO BIOTECHNOLOGIES INC Anti-Human CD3 (UCHT1) Purified, PE
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4073025
Encompass_Preferred
The Caprico PE Anti-Human CD3 Antibody is a phycoerythrin-conjugated mouse monoclonal antibody that recognizes the CD3 antigen on human T cells. It is intended for use in flow cytometry to identify and analyze T cell populations. The antibody has been tested and validated for flow cytometry applications. It is supplied as a liquid in PBS with 0.1% sodium azide and 1% BSA. According to the product details, this antibody clone has high sensitivity and specificity. The recommended usage is 5-10 μL per test, with typical dilutions ranging from 1:50 to 1:200. It is available in sizes of 100 tests and 500 tests.
Catalog No. 50-227-9220