Learn More
ACROBiosystems Recombinant Protein;Human CD58 / LFA-3 Protein, Fc Tag ;HEK293;1MG;LF3,CD58,LFA3,Ag3,LFA-3,Ag-3
Supplier:  ACROBiosystems LF3H52561MG
Encompass_Preferred
Recombinant Protein;Human CD58 / LFA-3 Protein, Fc Tag ;1MG;Human CD58, Fc Tag (LF3-H5256) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Arg 215 (Accession # AAH05930).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48.1 kDa. As a result of differtent glycosylation, the protein migrates as 60-75 kDa under reducing (R) condition, and 125-150 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human CD2, His Tag (Cat. No. CD2-H5226) at 2μg/mL (100 μL/well) can bind Human CD58, Fc Tag (Cat. No. LF3-H5256) with a linear range of 10-156 ng/mL (QC tested).
Catalog No. 50-165-8181