Learn More
ACROBiosystems Recombinant Protein;Human CD58 / LFA-3 Protein;HEK293;1MG;LF3,CD58,LFA3,Ag3,LFA-3,Ag-3
Supplier:  ACROBiosystems LF3H52251MG
Encompass_Preferred
Recombinant Protein;Human CD58 / LFA-3 Protein;1MG;Human CD58, His Tag (LF3-H5225) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Arg 215 (Accession # AAH05930).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.3 kDa. The protein migrates as 30-45 kDa under reducing (R) condition (SDS-PAGE) due to differtent glycosylation.
Catalog No. 50-165-8180