Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 966MSM4P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: MACIF/1193. Clonality: Monoclonal. Host: Mouse. Isotype: IgM kappa. Positive Control: Daudi, CEM, K562, HPB.ALL, Jurkat, Raji,, human lymphocytes. Human lymph node, tonsil. Cellular Staining: Cell Surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant full-length human CD59 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 20kDa.Chromosome: 11p13. Gene Symbol: CD59. Gene ID: 966. Ensemble: ENSG00000085063. Uniprot: NP_001120697.
Catalog No. 50-188-5367