Learn More
Biomatik Corporation Cd59 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC10357
Encompass_Preferred
Cd59 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to CD59. The Cd59 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Cd59 Glycoprotein Protein (26-102Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: CD59. Accession Number: P13987. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4329