Learn More
ACROBiosystems ACROBiosystems Human CD59 Protein, His Tag
Catalog No. 501956377
Encompass_Preferred
Recombinant Protein;1MG;Human CD59, His Tag (CD9-H52H9) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Asn 102 (Accession # P13987-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 10.9 kDa.
Catalog No. 50-195-6377 Supplier ACROBiosystems Supplier No. CD9H52H91MG
Recombinant Protein;1MG;Human CD59, His Tag (CD9-H52H9) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Asn 102 (Accession # P13987-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 10.9 kDa.