Learn More
Medchemexpress LLC CD59 protein, rat (HEK293, His) | >95.0% | 10 UG
SDP
Catalog No. 5000291755
Encompass_Preferred
Recombinant rat CD59 protein produced in HEK293 cells with a C-terminal His tag. CD59 is a membrane attack complex (MAC) inhibitor that regulates complement activation at or after the C5b-8 stage. Supplied as a purified recombinant protein for use in in vitro complement biology, functional assays, and antibody development. Available in microgram-scale quantities for laboratory research.
  • Recombinant rat CD59 expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • Greater than 95.0% purity as determined by reducing SDS-PAGE.
  • Available in microgram-scale quantities for laboratory assays.
  • Suitable for complement system studies and in vitro functional assays.
Catalog No. 50-002-91755 Supplier Medchemexpress LLC Supplier No. HYP7539010UG
Recombinant rat CD59 protein produced in HEK293 cells with a C-terminal His tag. CD59 is a membrane attack complex (MAC) inhibitor that regulates complement activation at or after the C5b-8 stage. Supplied as a purified recombinant protein for use in in vitro complement biology, functional assays, and antibody development. Available in microgram-scale quantities for laboratory research.
  • Recombinant rat CD59 expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • Greater than 95.0% purity as determined by reducing SDS-PAGE.
  • Available in microgram-scale quantities for laboratory assays.
  • Suitable for complement system studies and in vitro functional assays.