Learn More
ACROBiosystems Recombinant Protein;Human CD5 Protein;HEK293;100UG;CD5,CD5,LEU1,LEU-1
Catalog No. 501658749
Encompass_Preferred
Recombinant Protein;Human CD5 Protein;100UG;Human CD5, His Tag (CD5-H52H5) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Pro 372 (Accession # NP_055022).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40.5 kDa. The protein migrates as 48-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8749 Supplier ACROBiosystems Supplier No. CD5H52H5100UG
Recombinant Protein;Human CD5 Protein;100UG;Human CD5, His Tag (CD5-H52H5) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Pro 372 (Accession # NP_055022).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40.5 kDa. The protein migrates as 48-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.