Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 923MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: C6/372. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: CCRF-CEM, Jurkat cells, Tonsil. Cellular Staining: Cell surface, Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Human recombinant CD6 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 90-130kDa.Chromosome: 11q12.2. Gene Symbol: CD6. Gene ID: 923. Ensemble: ENSG00000013725. Uniprot: NP_001241679.
Catalog No. 50-188-5327